Assay Details
| Gene Symbol: PPCDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013973044.2 | XP_013828498.2 | 2 - 3 | 304 | 78 | |
| XM_013973045.2 | XP_013828499.2 | 2 - 3 | 284 | 78 | ||
| XM_013973046.2 | XP_013828500.2 | 2 - 3 | 232 | 78 | ||
| XM_013973047.2 | XP_013828501.2 | 2 - 3 | 292 | 78 | ||
| XM_018066372.1 | XP_017921861.1 | 2 - 3 | 284 | 78 | ||
| XM_018066374.1 | XP_017921863.1 | 2 - 3 | 284 | 78 | ||
| GenBank mRNA | JO595364.2 | - | 2 - 3 | 253 | 78 | |
More Information
Set Membership: |
|

PPCDC
Genomic Map