Assay Details
Gene Symbol: KDM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040743.1 | XP_017896232.1 | 3 - 4 | 801 | 59 | |
XM_018040744.1 | XP_017896233.1 | 3 - 4 | 801 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
