Assay Details
Gene Symbol: LHPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041189.1 | XP_017896678.1 | 4 - 5 | 631 | 57 | |
GenBank mRNA | JO433313.2 | - | 3 - 4 | 1021 | 57 | |
JO510598.2 | - | 4 - 5 | 562 | 57 | ||
KA326304.1 | - | 3 - 4 | 996 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
