Assay Details
| Gene Symbol: FAIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047815.1 | XP_017903304.1 | 11 - 12 | 948 | 55 | |
| GenBank mRNA | JO632007.2 | - | 11 - 12 | 1024 | 55 | |
More Information
Set Membership: |
|

FAIM2
Genomic Map