Assay Details
| Gene Symbol: CTDSPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053824.1 | XP_017909313.1 | 6 - 7 | 1208 | 69 | |
| XM_018053825.1 | XP_017909314.1 | 6 - 7 | 1236 | 69 | ||
| GenBank mRNA | JO600021.2 | - | 6 - 7 | 1034 | 69 | |
More Information
Set Membership: |
|

CTDSPL2
Genomic Map