Assay Details
Gene Symbol: BOLA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055022.1 | XP_017910511.1 | 3 - 4 | 63 | ||
GenBank mRNA | JO584651.1 | - | 3 - 4 | 389 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
