Assay Details
Gene Symbol: LOC102190533 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059837.1 | XP_017915326.1 | 16 - 17 | 3206 | 70 | |
XM_018059838.1 | XP_017915327.1 | 13 - 14 | 2858 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
