Assay Details
Gene Symbol: NDUFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691234.3 | XP_005691291.2 | 2 - 3 | 236 | 96 | |
GenBank mRNA | JO448491.2 | - | 2 - 3 | 233 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
