Assay Details
Gene Symbol: CREBRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005694535.3 | XP_005694592.1 | 5 - 6 | 1772 | 82 | |
XM_018065597.1 | XP_017921086.1 | 5 - 6 | 2295 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
