Assay Details
Gene Symbol: CIDEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695622.3 | XP_005695679.1 | 5 - 6 | 513 | 78 | |
XM_005695623.3 | XP_005695680.1 | 4 - 5 | 670 | 78 | ||
XM_018038446.1 | XP_017893935.1 | 4 - 5 | 559 | 78 | ||
XM_018038447.1 | XP_017893936.1 | 3 - 4 | 512 | 78 | ||
GenBank mRNA | JT750318.1 | - | 4 - 5 | 505 | 78 | |
KA322112.1 | - | 4 - 5 | 469 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
