Assay Details
| Gene Symbol: TADA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695627.3 | XP_005695684.1 | 8 - 9 | 1181 | 60 | |
| XM_018066924.1 | XP_017922413.1 | 8 - 9 | 1223 | 60 | ||
| XM_018066925.1 | XP_017922414.1 | 8 - 9 | 1132 | 60 | ||
| XM_018066926.1 | XP_017922415.1 | 8 - 9 | 1219 | 60 | ||
| XM_018066927.1 | XP_017922416.1 | 8 - 9 | 1233 | 60 | ||
| GenBank mRNA | JO576189.1 | - | 2 - 3 | 780 | 60 | |
More Information
Set Membership: |
|

TADA3
Genomic Map