Assay Details
| Gene Symbol: AGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699373.3 | XP_005699430.1 | 2 - 3 | 960 | 62 | |
| GenBank mRNA | JO421743.2 | - | 2 - 3 | 952 | 62 | |
| JO631757.2 | - | 3 - 4 | 1095 | 62 | ||
More Information
Set Membership: |
|

AGT
Genomic Map