Assay Details
Gene Symbol: WASHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048203.1 | XP_017903692.1 | 14 - 15 | 1412 | 106 | |
XM_018048204.1 | XP_017903693.1 | 11 - 12 | 1035 | 106 | ||
GenBank mRNA | JT666958.1 | - | 14 - 15 | 1397 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
