Assay Details
Gene Symbol: WASHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048203.1 | XP_017903692.1 | 20 - 21 | 2097 | 89 | |
XM_018048204.1 | XP_017903693.1 | 17 - 18 | 1720 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
