Assay Details
Gene Symbol: WASHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048203.1 | XP_017903692.1 | 22 - 23 | 2416 | 101 | |
XM_018048204.1 | XP_017903693.1 | 19 - 20 | 2039 | 101 | ||
GenBank mRNA | JT666958.1 | - | 22 - 23 | 2395 | 101 | |
JT689199.1 | - | 22 - 23 | 2370 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
