Assay Details
Gene Symbol: WASHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048203.1 | XP_017903692.1 | 30 - 31 | 3250 | 67 | |
XM_018048204.1 | XP_017903693.1 | 27 - 28 | 2873 | 67 | ||
GenBank mRNA | JT666958.1 | - | 30 - 31 | 3238 | 67 | |
JT689199.1 | - | 30 - 31 | 3207 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
