Assay Details
| Gene Symbol: PCDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050216.1 | XP_017905705.1 | 2 - 3 | 1068 | 67 | |
| XM_018050217.1 | XP_017905706.1 | 2 - 3 | 957 | 67 | ||
| XM_018050218.1 | XP_017905707.1 | 2 - 3 | 1068 | 67 | ||
| GenBank mRNA | JT655581.1 | - | 7 - 8 | 3742 | 67 | |
More Information
Set Membership: |
|

PCDH1
Genomic Map