Assay Details
Gene Symbol: RAB11FIP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055043.1 | XP_017910532.1 | 5 - 6 | 3950 | 86 | |
XM_018055044.1 | XP_017910533.1 | 5 - 6 | 3949 | 86 | ||
XM_018055045.1 | XP_017910534.1 | 5 - 6 | 3652 | 86 | ||
XM_018055046.1 | XP_017910535.1 | 5 - 6 | 3191 | 86 | ||
XM_018055047.1 | XP_017910536.1 | 4 - 5 | 1942 | 86 | ||
XM_018055048.1 | XP_017910537.1 | 4 - 5 | 1991 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
