Assay Details
Gene Symbol: CCDC62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013970643.2 | XP_013826097.2 | 11 - 12 | 2124 | 72 | |
XM_018061331.1 | XP_017916820.1 | 12 - 13 | 2283 | 72 | ||
XM_018061332.1 | XP_017916821.1 | 12 - 13 | 2280 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
