Assay Details
Gene Symbol: MMP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064218.1 | XP_017919707.1 | 2 - 3 | 427 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
