Assay Details
| Gene Symbol: IFI6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056827.1 | XP_017912316.1 | 2 - 3 | 214 | 75 | |
| XM_018056828.1 | XP_017912317.1 | 3 - 4 | 282 | 75 | ||
| GenBank mRNA | JO632972.2 | - | 3 - 4 | 480 | 75 | |
| JR129141.1 | - | 939 | 75 | |||
More Information
Set Membership: |
|

IFI6
Genomic Map