Assay Details
Gene Symbol: TEDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040988.1 | XP_017896477.1 | 3 - 4 | 252 | 83 | |
XM_018040989.1 | XP_017896478.1 | 3 - 4 | 252 | 83 | ||
XM_018040990.1 | XP_017896479.1 | 3 - 4 | 252 | 83 | ||
XM_018040991.1 | XP_017896480.1 | 3 - 4 | 252 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
