Assay Details
| Gene Symbol: FOLH1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043532.1 | XP_017899021.1 | 10 - 11 | 1347 | 70 | |
| GenBank mRNA | JT730147.1 | - | 12 - 13 | 1367 | 70 | |
More Information
Set Membership: |
|

FOLH1B
Genomic Map