Assay Details
| Gene Symbol: SLC25A29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066354.1 | XP_017921843.1 | 1 - 2 | 1939 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC25A29| Gene Symbol: SLC25A29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066354.1 | XP_017921843.1 | 1 - 2 | 1939 | 128 | |
Set Membership: |
|