Assay Details
| Gene Symbol: ORAI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697716.3 | XP_005697773.3 | 1 - 2 | 797 | 79 | |
| GenBank mRNA | JO356342.2 | - | 1 - 2 | 144 | 79 | |
| JO578501.1 | - | 1 - 2 | 498 | 79 | ||
More Information
Set Membership: |
|

ORAI3
Genomic Map