Assay Details
| Gene Symbol: ACTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063603.1 | XP_017919092.1 | 3 - 4 | 126 | ||
| GenBank mRNA | JT712426.1 | - | 1 | 1426 | 233 | |
More Information
Set Membership: |
|

ACTG1
Genomic Map