Assay Details
Gene Symbol: TRA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047312.1 | XP_017902801.1 | 8 | 1964 | 88 | |
XM_018047313.1 | XP_017902802.1 | 8 | 2554 | 88 | ||
XM_018047314.1 | XP_017902803.1 | 9 | 2644 | 88 | ||
XM_018047315.1 | XP_017902804.1 | 8 | 3692 | 88 | ||
XM_018047316.1 | XP_017902805.1 | 9 | 2274 | 88 | ||
XM_018047317.1 | XP_017902806.1 | 9 | 2057 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
