Assay Details
Gene Symbol: PTRH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693127.2 | XP_005693184.1 | 2 | 723 | 122 | |
XM_005693128.3 | XP_005693185.1 | 3 | 808 | 122 | ||
GenBank mRNA | JO348186.2 | - | 2 | 623 | 122 | |
JO348188.2 | - | 1 | 1116 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
