Assay Details
| Gene Symbol: TSHZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039574.1 | XP_017895063.1 | 2 | 3139 | 102 | |
| GenBank mRNA | JT669987.1 | - | 4 | 1377 | 102 | |
More Information
Set Membership: |
|

TSHZ1
Genomic Map