Assay Details
Gene Symbol: ARL4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693897.2 | XP_005693954.1 | 2 | 283 | 89 | |
GenBank mRNA | JO439380.2 | - | 2 | 238 | 89 | |
KA306793.1 | - | 1 | 1079 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
