Assay Details
| Gene Symbol: POLR2K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689201.2 | XP_005689258.1 | 4 | 1025 | 94 | |
| XM_018058301.1 | XP_017913790.1 | 4 | 1161 | 94 | ||
| XM_018058302.1 | XP_017913791.1 | 4 | 1035 | 94 | ||
More Information
Set Membership: |
|

POLR2K
Genomic Map