Assay Details
| Gene Symbol: PTGES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055956.1 | XP_017911445.1 | 3 | 453 | 96 | |
| GenBank mRNA | KA351519.1 | - | 3 | 419 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PTGES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055956.1 | XP_017911445.1 | 3 | 453 | 96 | |
| GenBank mRNA | KA351519.1 | - | 3 | 419 | 96 | |
Set Membership: |
|