Assay Details
Gene Symbol: YWHAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060921.1 | XP_017916410.1 | 2 | 432 | 115 | |
GenBank mRNA | JO633596.2 | - | 1 | 1336 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
