Assay Details
Gene Symbol: ZNF608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744648.3 | XP_002744694.1 | 2 - 3 | 1674 | 61 | |
XM_008991602.2 | XP_008989850.1 | 2 - 3 | 1380 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
