Assay Details
| Gene Symbol: PPM1K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745604.3 | XP_002745650.1 | 5 - 6 | 1333 | 70 | |
| XM_008992999.2 | XP_008991247.1 | 7 - 8 | 1399 | 70 | ||
| XM_008993000.2 | XP_008991248.1 | 7 - 8 | 1107 | 70 | ||
| XM_008993001.2 | XP_008991249.1 | 7 - 8 | 1138 | 70 | ||
More Information
Set Membership: |
|

PPM1K
Genomic Map