Assay Details
| Gene Symbol: AP3S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744690.4 | XP_002744736.1 | 1 - 2 | 504 | 65 | |
| XM_008991658.2 | XP_008989906.1 | 1 - 2 | 504 | 65 | ||
| XM_008991659.2 | XP_008989907.1 | 1 - 2 | 504 | 65 | ||
| XM_017969730.1 | XP_017825219.1 | 1 - 2 | 212 | 65 | ||
| XM_017969731.1 | XP_017825220.1 | 1 - 2 | 212 | 65 | ||
| XM_017969732.1 | XP_017825221.1 | 1 - 2 | 468 | 65 | ||
More Information
Set Membership: |
|

AP3S1
Genomic Map