Assay Details
Gene Symbol: YWHAE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002747823.4 | XP_002747869.1 | 2 - 3 | 508 | 106 | |
XM_008996385.2 | XP_008994633.1 | 2 - 3 | 395 | 106 | ||
XM_008996386.1 | XP_008994634.1 | 3 - 4 | 442 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
