Assay Details
| Gene Symbol: CENPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993252.2 | XP_008991500.1 | 16 - 17 | 2736 | 96 | |
| XM_017970511.1 | XP_017826000.1 | 15 - 16 | 2676 | 96 | ||
More Information
Set Membership: |
|

CENPC
Genomic Map