Assay Details
| Gene Symbol: SEC22A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734908.3 | XP_003734956.1 | 5 - 6 | 639 | 67 | |
| XM_008982326.2 | XP_008980574.1 | 5 - 6 | 978 | 67 | ||
| XM_008982328.2 | XP_008980576.1 | 5 - 6 | 701 | 67 | ||
| XM_008982329.2 | XP_008980577.1 | 5 - 6 | 1031 | 67 | ||
| XM_008982332.2 | XP_008980580.1 | 5 - 6 | 654 | 67 | ||
| XM_017965183.1 | XP_017820672.1 | 5 - 6 | 664 | 67 | ||
More Information
Set Membership: |
|

SEC22A
Genomic Map