Assay Details
| Gene Symbol: MSS51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009713.2 | XP_009007961.1 | 5 - 6 | 1417 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MSS51| Gene Symbol: MSS51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009713.2 | XP_009007961.1 | 5 - 6 | 1417 | 85 | |
Set Membership: |
|