Assay Details
Gene Symbol: LOC100394842 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008997150.2 | XP_008995398.1 | 4 - 5 | 574 | 69 | |
XM_017972558.1 | XP_017828047.1 | 5 - 6 | 1809 | 69 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
