Assay Details
| Gene Symbol: ARHGAP22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978921.1 | XP_017834410.1 | 6 - 7 | 1183 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARHGAP22| Gene Symbol: ARHGAP22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978921.1 | XP_017834410.1 | 6 - 7 | 1183 | 66 | |
Set Membership: |
|