Assay Details
Gene Symbol: LMX1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008985113.2 | XP_008983361.1 | 7 - 8 | 1175 | 64 | |
XM_008985114.2 | XP_008983362.1 | 6 - 7 | 996 | 64 | ||
XM_017966120.1 | XP_017821609.1 | 6 - 7 | 1315 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
