Assay Details
Gene Symbol: ARCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009006926.2 | XP_009005174.2 | 2 - 3 | 410 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
