Assay Details
| Gene Symbol: EIF4E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745571.4 | XP_002745617.1 | 4 - 5 | 1163 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EIF4E| Gene Symbol: EIF4E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745571.4 | XP_002745617.1 | 4 - 5 | 1163 | 80 | |
Set Membership: |
|