Assay Details
Gene Symbol: GPBP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002750771.4 | XP_002750817.1 | 4 - 5 | 1530 | 99 | |
XM_009001136.2 | XP_008999384.1 | 4 - 5 | 1530 | 99 | ||
XM_017974674.1 | XP_017830163.1 | 4 - 5 | 1530 | 99 | ||
XM_017974675.1 | XP_017830164.1 | 4 - 5 | 1456 | 99 | ||
XM_017974676.1 | XP_017830165.1 | 4 - 5 | 1558 | 99 | ||
XM_017974677.1 | XP_017830166.1 | 4 - 5 | 1499 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
