Assay Details
| Gene Symbol: KCTD13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756047.4 | XP_002756093.1 | 4 - 5 | 766 | 87 | |
| XM_017978853.1 | XP_017834342.1 | 5 - 6 | 829 | 87 | ||
| XM_017978854.1 | XP_017834343.1 | 5 - 6 | 829 | 87 | ||
More Information
Set Membership: |
|

KCTD13
Genomic Map