Assay Details
| Gene Symbol: RCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742905.4 | XP_002742951.1 | 6 - 7 | 972 | 102 | |
| XM_017973412.1 | XP_017828901.1 | 6 - 7 | 967 | 102 | ||
| XM_017973414.1 | XP_017828903.1 | 6 - 7 | 967 | 102 | ||
More Information
Set Membership: |
|

RCL1
Genomic Map