Assay Details
| Gene Symbol: ECHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746939.4 | XP_002746985.1 | 5 - 6 | 975 | 85 | |
| XM_008994998.2 | XP_008993246.1 | 6 - 7 | 1150 | 85 | ||
| XM_008995000.2 | XP_008993248.1 | 5 - 6 | 1097 | 85 | ||
| XM_008995001.2 | XP_008993249.1 | 5 - 6 | 1140 | 85 | ||
| XM_017971377.1 | XP_017826866.1 | 6 - 7 | 596 | 85 | ||
More Information
Set Membership: |
|

ECHDC1
Genomic Map