Assay Details
Gene Symbol: MYLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017966863.1 | XP_017822352.1 | 12 - 13 | 2207 | 63 | |
XM_017966864.1 | XP_017822353.1 | 12 - 13 | 3980 | 63 | ||
XM_017966865.1 | XP_017822354.1 | 12 - 13 | 2196 | 63 | ||
XM_017966866.1 | XP_017822355.1 | 12 - 13 | 2139 | 63 | ||
XM_017966867.1 | XP_017822356.1 | 12 - 13 | 2192 | 63 | ||
XM_017966868.1 | XP_017822357.1 | 12 - 13 | 2126 | 63 | ||
XM_017966869.1 | XP_017822358.1 | 10 - 11 | 1567 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
